PIG® medium and large mobile container kits
Spill kit, Oil-Only, includes 160 mat pads, 14 socks, 18 pillows and 15 disposal bags, 250,8 L, 850×620×1100 mm
Catalog # 121-0073
Supplier: NEW PIG
Wechseln zu:
Product Details & Documents
Fast, effective response to indoor and outdoor spills.
- Durable, wheeled 240 and 360 L container is packed with high-quality PIG® absorbents
- Easy to transport supplies directly to the spill site
- Hinged lid makes it easy to get the absorbents you need — just flip it open
- Container with bold Spill Kit lettering for quick identification
- Absorbents are organised for quick, easy response
- Heavy-duty rubber wheels roll easily over nearly any surface
Weather-resistant polyethylene container protects the contents from moisture, dirt and damage (containers are best kept indoors for long-term storage).
Achtung! Bitte beachten: To ensure effectiveness and your safety, we recommend that you conduct compatibility and absorption testing of your chemicals with PIG® HAZ-MAT products prior to purchase.
Spezifikationen:
- Description:Spill kit, Oil-Only, includes 160 mat pads, 14 socks, 18 pillows and 15 disposal bags
- Seq:11
- W×D×H:850×620×1100 mm
- Absorbency:250,8 L
Empfehlungen
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen.



