Orders LinkContactUsLinkComponent
Multispectral source, BioLite™
Multispectral source, BioLite™
Manual BioLite™ unit for in vivo studies, with EPI fibre optics, for darkroom with top access port*
Catalog # UVPA98-0072-11
Multispectral source, BioLite™
Catalog # UVPA98-0072-11
Multispectral source, BioLite™
Multispectral source, BioLite™
Manual BioLite™ unit for in vivo studies, with EPI fibre optics, for darkroom with top access port*
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
The BioLite MultiSpectral Source uses a 150 W quartz halogen or 150 W xenon light source with fibre optics to tightly control the output spectrum.
  • Xenon light source is strongly recommended for customers with NIR requirement
A six position dial allows selection of the light intensity output for different applications.
* No excitation filters are included with the BioLite unit. Please ask for details of the available choice.
Specifications
  • Description:
    Manual BioLite™ unit for in vivo studies, with EPI fibre optics, for darkroom with top access port*
  • Seq:
    42