Multispectral source, BioLite™
Automated BioLite™ unit for in vivo studies, with Epi fibre optics, for darkroom with top access port*
Catalog # UVPA98-0072-08
Supplier: UVP ULTRA VIOLET PRODUCTS
Jump to:
Product Details & Documents
The BioLite MultiSpectral Source uses a 150 W quartz halogen or 150 W xenon light source with fibre optics to tightly control the output spectrum.
- Xenon light source is strongly recommended for customers with NIR requirement
A six position dial allows selection of the light intensity output for different applications.
* No excitation filters are included with the BioLite unit. Please ask for details of the available choice.
Specifications
- Description:Automated BioLite™ unit for in vivo studies, with Epi fibre optics, for darkroom with top access port*
- Seq:41