Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Lanolin

Glass bottle for solids, 1 kg
Catalog # CSLO3024.1000
Marketsource
Supplier:   CAESAR LORENTZ
Lanolin
Catalog # CSLO3024.1000
Supplier:  CAESAR LORENTZ
Marketsource
CAS Number:  8006-54-0
Lanolin

Glass bottle for solids, 1 kg

CAS Number:  8006-54-0
Price
71,30 €
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  Wool fat
  • CAS Number:8006-54-0
  • MDL Number:MFCD00081740
  • Merck Index:12,05371
  • Melting Pt:38…44 °C
  • Density:0.932…0.945 g/cm³ (15 °C)
  • Flash Pt:113 °C (closed cup)
  • EINECS:232-348-6
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg

Specifications