Universal absorbent pillows
Universal pillow, 20 pillows/pack, 250×250 mm
Catalog # MSGA9944
Supplier: asecos
Jump to:
Product Details & Documents
Universal pillows for safe absorption of oil, water, cooling liquids and non aggressive chemicals.
- Optimal for those hard to reach places
- Capacity up to 16 times of its own weight
- High rib and rub resistance
Specifications
- Description:Universal pillow
- L×W:250×250 mm
- Packed:20 pillows/pack