Universal mats
Universal mat, standard, 200 mats/pack, 400×460 mm
Catalog # MSGA11326
Supplier: asecos
Jump to:
Product Details & Documents
Universal mats for safe absorption of oil, water, cooling liquids and non-aggressive chemicals.
- Capacity up to 16 times of its own weight
- High rib and rub resistance
- Mats are pre-cut: only take sufficient material as required
Specifications
- Description:Universal mat, standard
- L×W:400×460 mm
- Packed:200 mats/pack