Order Entry
Germany
Orders LinkContactUsLinkComponent
6-Hydroxyindole 95%

6-Hydroxyindole 95%

Bottle, 5 g
Catalog # APOSOR7612-5G
Corrosive
Irritant
Environment
Supplier:   Apollo Scientific
6-Hydroxyindole 95%
Catalog # APOSOR7612-5G
Supplier:  Apollo Scientific
CAS Number:  2380-86-1
6-Hydroxyindole 95%

Bottle, 5 g

CAS Number:  2380-86-1
Price
189,00 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  Indol-6-ol, 6-Indolol, IMEXINE OBA, 6-Hydroxy-1H-indole, 1H-Indol-6-ol
  • CAS Number:2380-86-1
  • Formula:C₈H₇NO
  • MDL Number:MFCD00152101
  • Molecular Weight:133.15 g/mol
  • Melting Pt:125…128 °C
  • Storage Temperature:Refrigerator
  • EINECS:417-020-4
  • UN:3077
  • ADR:9+EHS,III
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.