Orders LinkContactUsLinkComponent
Safety boots, pull on, Jalfrigg
Safety boots, pull on, Jalfrigg
44, Beige
Catalog # 114-4022
Supplier:   Jallatte
Safety boots, pull on, Jalfrigg
Catalog # 114-4022
Supplier:  Jallatte
Safety boots, pull on, Jalfrigg
Safety boots, pull on, Jalfrigg
44, Beige
Price
Inquire For Price
Log in for availability
1 * 1 PAAR
Product Details & Documents
Metal-free safety boots in natural full-grain leather, highly water and oil resistant.
  • Softane™ dual-density PU provides excellent absorption of heel shocks and vibrations
  • Lightweight, non magnetic, perforated composite toecap (200 J)
  • Non magnetic, lightweight and flexible FleXtane™ HP midsole (100% composite)
Fully lined for use in cold environments.
Colour: Beige
Certifications: EN ISO 20345; S3, CI, SRC
Specifications
  • Shoe size:
    44
  • Colour:
    Beige