ContactUsLinkComponent
Ammonia combination electrode with screw plug head, Plastic shaft, exchangable membrane modul, measuring range 0.1 mg/l...1000 mg/l, 0...+50 °C, screw plug head, length 120 mm, 12 mm Ø 1 * 1 items
Catalog # SCOI1064386
Supplier:  SI Analytics
Ammonia combination electrode with screw plug head, Plastic shaft, exchangable membrane modul, measuring range 0.1 mg/l...1000 mg/l, 0...+50 °C, screw plug head, length 120 mm, 12 mm Ø 1 * 1 items
Catalog # SCOI1064386
Supplier:  SI Analytics
Frequently Bought Together