Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Labelling tapes

Labelling tapes, Green, 25,0 mm
  SEMA4340
 :   SEMADENI
Labelling tapes
  SEMA4340
 :  SEMADENI
Labelling tapes
Labelling tapes

Labelling tapes, Green, 25,0 mm

 
15,20 €
   
1 * 1 ST
 

Labeling tape for glass, plastic, porcelain or metal.

  • Tear resistant
  • Can be easily written on
  • Adhesive strength at low temperature

Length: 66,0 m

Different colours available.

 : Do not autoclave.

 
  • Width:
    25,0 mm
  • Colour:
    Green
  • Description:
    Labelling tapes
  • Seq:
    06
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.