Order Entry
Germany
Orders LinkContactUsLinkComponent
Slide box

Slide box

Dust cover lids, 345×35×40 mm
Catalog # 630-1323
Supplier:   KARTELL
Slide box
Catalog # 630-1323
Supplier:  KARTELL
Slide box
Slide box

Dust cover lids, 345×35×40 mm

Price
48,90 €
Log in for availability
1 * 2 ST
Product Details & Documents

Each cavity is numbered for storage in drawers.

W×D×H: 38×360×100 mm

Specifications
  • Description:
    Dust cover lids
  • W×D×H:
    345×35×40 mm