Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Storage bin for MF22/30 iceflakers 85kg capacity 1 * 1 items
Catalog # SCOMB190S
Supplier:
SCOTSMAN
Expand image
Storage bin for MF22/30 iceflakers 85kg capacity 1 * 1 items
Catalog # SCOMB190S
Supplier:
SCOTSMAN