Order Entry
Germany
Orders LinkContactUsLinkComponent
HPLC columns, SeQuant® ZIC®-HILIC | Avantor

HPLC columns, SeQuant® ZIC®-HILIC

ZIC®-HILIC HPLC column, Stainless steel, PEEK coated, 200 Å, 5 µm, 2,10 mm, 100 mm
Catalog # 1.50452.0001
Supplier:   Merck
HPLC columns, SeQuant® ZIC®-HILIC
Catalog # 1.50452.0001
Supplier:  Merck
HPLC columns, SeQuant® ZIC®-HILIC
HPLC columns, SeQuant® ZIC®-HILIC

ZIC®-HILIC HPLC column, Stainless steel, PEEK coated, 200 Å, 5 µm, 2,10 mm, 100 mm

Wechseln zu:
Price
1 060,00 €
Anmelden für die Verfügbarkeit
1 * 1 ST
Product Details & Documents

The selectivity offered by ZIC®-HILIC is suitable for a wide variety of molecules containing hydrophilic or ionisable functional groups. This includes compounds such as carbohydrates, metabolites, acids and bases, organic and inorganic ions, metal complexes, amino acids, peptides, protein digests, plant and cell extracts, plus much more. Such compounds are normally characterised by a small or negative LogP value and have poor retention on reversed-phase columns.

  • Straightforward separation of polar and hydrophilic compounds
  • Orthogonal selectivity compared to reversed-phase
  • High density-bonded sulphobetaine zwitterionic stationary phase
  • Low cost per analysis due to robust stationary phase

* HILIC = Hydrophilic Interaction Liquid Chromatography. *LogP = Octanol-water partition coefficient.

Spezifikationen:
  • Ø int.:
    2,10 mm
  • Particle Size:
    5 µm
  • Pore size:
    200 Å
  • Description:
    ZIC®-HILIC HPLC column
  • Length:
    100 mm
  • Seq:
    13
  • Column hardware:
    Stainless steel, PEEK coated
Empfehlungen

Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.

Andernfalls erhalten Sie allgemeine Empfehlungen.