Order Entry
Germany
Orders LinkContactUsLinkComponent
Spare keys for stopcocks, Pyrex®

Spare keys for stopcocks, Pyrex®

Replacement keys, for 14,5 mm Ø, 4,0 mm, SSD series stopcocks
  BIBBSSD104K
 :   SciLabware
Spare keys for stopcocks, Pyrex®
  BIBBSSD104K
Spare keys for stopcocks, Pyrex®
Spare keys for stopcocks, Pyrex®

Replacement keys, for 14,5 mm Ø, 4,0 mm, SSD series stopcocks

 
26,20 €
   
1 * 1 ST
 

Spare keys for stopckocks, made of borosilicate glass.

  • Straight bore
  • Interchangeable

Delivery information: Supplied complete with key retaining device.

 
  • For:
    SSD series stopcocks
  • Hole Ø:
    4,0 mm
  • Description:
    Replacement keys, for 14,5 mm Ø
  • Seq:
    02
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.