Orders LinkContactUsLinkComponent
Electrode Cables
Electrode Cables
Electrode cable, with plug H, for connecting generator electrode - KF Coulometer, 1 m
Catalog # METR6.2104.120
Supplier:   METROHM
Electrode Cables
Catalog # METR6.2104.120
Supplier:  METROHM
Electrode Cables
Electrode cable, with plug H, for connecting generator electrode - KF Coulometer, 1 m
Price
Inquire For Price
Log in for availability
1 * 1 m
Product Details & Documents
Specifications
  • Description:
    Electrode cable, with plug H, for connecting generator electrode - KF Coulometer
  • Length:
    1 m
  • Seq:
    07