ContactUsLinkComponent
Salmonella enterica 10 ml Growth Promotion Suspensions, USP/EP compliant, provide a QC challenge to each lot of medium whether purch ased commerically or prepared in-house. Population level of <100 Colony Forming Units (CFUs) per each 0.1 ml dose. 1 * 10 ml
Catalog # NAMSGP-11
Supplier:  CROSSTEX
Salmonella enterica 10 ml Growth Promotion Suspensions, USP/EP compliant, provide a QC challenge to each lot of medium whether purch ased commerically or prepared in-house. Population level of <100 Colony Forming Units (CFUs) per each 0.1 ml dose. 1 * 10 ml
Catalog # NAMSGP-11
Supplier:  CROSSTEX