Order Entry
Germany
Orders LinkContactUsLinkComponent
Spore strips

Spore strips

Bacillus atrophaeus cell line 9372: mini strip 106 (24 month shelf life) / sterilisation process: EO, 10 mm, 2 mm
Catalog # NAMSSTN-062B
Supplier:   CROSSTEX
Spore strips
Catalog # NAMSSTN-062B
Supplier:  CROSSTEX
Spore strips

Bacillus atrophaeus cell line 9372: mini strip 106 (24 month shelf life) / sterilisation process: EO, 10 mm, 2 mm

Price
246,00 €
Log in for availability
1 * 100 ST
Product Details & Documents

Documents

Specifications
  • Width:
    2 mm
  • Description:
    Bacillus atrophaeus cell line 9372: mini strip 106 (24 month shelf life) / sterilisation process: EO
  • Length:
    10 mm
  • Seq:
    01
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.