Order Entry
Germany
Orders LinkContactUsLinkComponent
Human Recombinant FGF2 (from E. coli)

Human Recombinant FGF2 (from E. coli)

Catalog # ABCAAB61845-50
Marketsource
Supplier:   Abcam
Human Recombinant FGF2 (from E. coli)
Catalog # ABCAAB61845-50
Supplier:  Abcam
Marketsource
CAS Number:  
Human Recombinant FGF2 (from E. coli)
Human Recombinant FGF2 (from E. coli)
CAS Number:  
Price
344,00 €
Log in for availability
1 * 50 µG
Product Details & Documents

Recombinant human FGF2 protein is a Human Full Length protein, expressed in Escherichia coli, with >95% purity and suitable for SDS-PAGE, FuncS.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifications

Specifications

  • Form:
    Lyophilised
  • Protein/peptide name:
    FGF2
  • Protein synonyms:
    FGFB|Fibroblast growth factor 2|FGF-2|Basic fibroblast growth factor|Heparin-binding growth factor 2|bFGF|HBGF-2
  • Purity:
    >95%
  • Source:
    E. coli
  • Biological activity:
    Standard
  • Shipping temperature:
    20
  • Cat. No.:
    ABCAAB61845-50
  • Protein/peptide type:
    Recombinant
  • Storage conditions:
    Store at −20 °C
  • Formulation:
    Constituents: 0,164% Sodium phosphate
  • Conjugation:
    Unconjugated
  • Tested applications:
    FuncS, SDS-PAGE
  • Species:
    Human
  • Animal-free:
    N

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.