Orders LinkContactUsLinkComponent
Replacement plungers
Replacement plungers
Replacement plunger, 25 µl X-type syringes
Catalog # HEWLG4200-80112
Supplier:   Agilent
Replacement plungers
Catalog # HEWLG4200-80112
Supplier:  Agilent
Replacement plungers
Replacement plungers
Replacement plunger, 25 µl X-type syringes
Price
Inquire For Price
Log in for availability
1 * 3 ST
Product Details & Documents
Replacement plungers designed for long-lasting applications that demand precision and productivity.
  • Strongly recommended for high-throughput applications
  • X-type syringes feature zero carryover
Specifications
  • For:
    25 µl X-type syringes
  • Description:
    Replacement plunger
  • Seq:
    06