Replacement plungers
Replacement plunger, 100 µl X-type syringes
Catalog # HEWLG4200-80111
Supplier: Agilent
Jump to:
Product Details & Documents
Replacement plungers designed for long-lasting applications that demand precision and productivity.
- Strongly recommended for high-throughput applications
- X-type syringes feature zero carryover
Specifications
- For:100 µl X-type syringes
- Description:Replacement plunger
- Seq:07