Order Entry
Germany
Orders LinkContactUsLinkComponent
Capillaries for LC systems

Capillaries for LC systems

Capillaries, pre-swaged, red, stainless steel, 210 mm, I.Ø 0,12 mm
Catalog # HEWLG1316-87328
Supplier:   Agilent
Capillaries for LC systems
Catalog # HEWLG1316-87328
Supplier:  Agilent
Capillaries for LC systems
Capillaries for LC systems

Capillaries, pre-swaged, red, stainless steel, 210 mm, I.Ø 0,12 mm

Price
102,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Stainless steel capillaries are made of flexible stainless steel (0,6 mm OD) with ¹/₁₆" OD tubing at both ends to accept standard fittings.

Pre-swaged fittings are assembled according to Swagelok specifications.

Specifications
  • Description:
    Capillaries, pre-swaged, red, stainless steel, 210 mm, I.Ø 0,12 mm
  • Seq:
    60