cis-4-Hydroxy-L-(-)-proline ≥99%
Glass bottle for solids, 1 g
Catalog # H27194.03
Supplier: Thermo Fisher Scientific
cis-4-Hydroxy-L-(-)-proline ≥99%
Catalog # H27194.03
Supplier: Thermo Fisher Scientific
CAS Number: 618-27-9
Jump to:
Product Details & Documents
Purity: 99% CAS No.: 618-27-9 Molecular Formula: C5H9NO3 Formula Weight: 131.13 Melting Point: ca 250° dec. Optical Rotation: -59° (c=2 in water)
Synonyms: L(-)-cis-4-Hydroxyproline, H-cis-Hyp-OH, (2S,4S)-4-Hydroxypyrrolidine-2-carboxylic acid, (S)-(-)-cis-4-Hydroxyproline
- CAS Number:618-27-9
- Formula:C₅H₉NO₃
- MDL Number:MFCD00064319
- Molecular Weight:131.13 g/mol
- Melting Pt:∼ 250 °C (dec.)
- Storage Temperature:Ambient
- EINECS:210-542-1
Documents
Organic Building Blocks and Intermediates
Application Notes
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



