Order Entry
Germany
Orders LinkContactUsLinkComponent
cis-4-Hydroxy-L-(-)-proline ≥99%

cis-4-Hydroxy-L-(-)-proline ≥99%

Glass bottle for solids, 250 mg
Catalog # H27194.MD
cis-4-Hydroxy-L-(-)-proline ≥99%
Catalog # H27194.MD
CAS Number:  618-27-9
cis-4-Hydroxy-L-(-)-proline ≥99%
cis-4-Hydroxy-L-(-)-proline ≥99%

Glass bottle for solids, 250 mg

CAS Number:  618-27-9
Price
222,00 €
Log in for availability
1 * 250 mg
Product Details & Documents

Purity: 99% CAS No.: 618-27-9 Molecular Formula: C5H9NO3 Formula Weight: 131.13 Melting Point: ca 250° dec. Optical Rotation: -59° (c=2 in water)

Synonyms:  L(-)-cis-4-Hydroxyproline, H-cis-Hyp-OH, (2S,4S)-4-Hydroxypyrrolidine-2-carboxylic acid, (S)-(-)-cis-4-Hydroxyproline
  • CAS Number:618-27-9
  • Formula:C₅H₉NO₃
  • MDL Number:MFCD00064319
  • Molecular Weight:131.13 g/mol
  • Melting Pt:∼ 250 °C (dec.)
  • Storage Temperature:Ambient
  • EINECS:210-542-1
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.