Order Entry
Germany
Orders LinkContactUsLinkComponent
Cyclohexane for HPLC

Cyclohexane for HPLC

Glass bottle, 2.500 mL
Catalog # H26081.0F
Flammable
Health
Irritant
Environment
Cyclohexane for HPLC
Catalog # H26081.0F
CAS Number:  110-82-7
Cyclohexane for HPLC
Cyclohexane for HPLC

Glass bottle, 2.500 mL

CAS Number:  110-82-7
Price
193,00 €
Log in for availability
1 * 2.500 mL
Product Details & Documents
Synonyms:  Hexahydrobenzene, Hexamethylene
  • CAS Number:110-82-7
  • Formula:C₆H₁₂
  • MDL Number:MFCD00003814
  • Molecular Weight:84.16 g/mol
  • Merck Index:13,02752
  • Boiling Pt:81 °C (1013 hPa)
  • Melting Pt:7 °C
  • Density:0.7785 g/cm³ (20 °C)
  • Flash Pt:–18 °C
  • Storage Temperature:Ambient
  • EINECS:203-806-2
  • UN:1145
  • ADR:3,II
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2.500 mL

Specifications