Orders LinkContactUsLinkComponent
Counting chamber, McMaster
Counting chamber, McMaster
Cover glass with three counting grids (10×10 mm) divided into 10 parts
Catalog # MARI0335001
Supplier:   MARIENFELD
Counting chamber, McMaster
Catalog # MARI0335001
Supplier:  MARIENFELD
Counting chamber, McMaster
Counting chamber, McMaster
Cover glass with three counting grids (10×10 mm) divided into 10 parts
Price
131,53 €
Log in for availability
1 * 1 ST
Product Details & Documents
This cell counter is suitable for liquid media.
  • Three counting grids and spare cover glass
  • Pair of clamps to fix the cover glass
  • Ground edges and rounded corners
Specifications
  • Description:
    Cover glass with three counting grids (10×10 mm) divided into 10 parts