Brij® 35
Glass bottle for solids, 1.000 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
CAS No.: 9002-92-0 Molecular Formula: C58H118O24 Formula Weight: 1199.57 Physical Form: Waxy solid Appearance: White Melting Point: ca 35-40° MDL No.: MFCD00080891
For use in Stein-Moore chromatography
Synonyms: Polyoxyethylene(23)lauryl ether, Tricosaethylene glycol mono-n-dodecyl ether, Brij® L23
- CAS Number:9002-92-0
- Formula:C₅₈H₁₁₈O₂₄
- MDL Number:MFCD00080891
- Molecular Weight:1199.56 g/mol
- Melting Pt:∼ 35…40 °C
- Storage Temperature:Ambient
- EINECS:500-002-6
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1.000 g



