Order Entry
Germany
Orders LinkContactUsLinkComponent
ADA ≥98%

ADA ≥98%

Glass bottle for solids, 500 g
Catalog # A15267.36
ADA ≥98%
Catalog # A15267.36
CAS Number:  26239-55-4
ADA ≥98%
ADA ≥98%

Glass bottle for solids, 500 g

CAS Number:  26239-55-4
Price
470,00 €
Log in for availability
1 * 500 g
Product Details & Documents

Purity: 98+% CAS No.: 26239-55-4 Molecular Formula: H2NCOCH2N(CH2CO2H)2 Formula Weight: 190.16 Melting Point: ca 218° dec. Beilstein Reference: 1787181 Merck Reference: 14,147 MDL No.: MFCD00008031

Synonyms:  2,2'-[(2-Amino-2-oxoethyl)imino]diacetic acid, N-(2-Acetamido)iminodiacetic acid, 2,2'-((2-Amino-2-oxoethyl)azanediyl)diacetic acid
  • CAS Number:26239-55-4
  • Formula:H₂NC(O)CH₂N(CH₂COOH)₂
  • MDL Number:MFCD00008031
  • Molecular Weight:190.16 g/mol
  • Merck Index:14,147
  • Melting Pt:∼ 218 °C (dec.)
  • Storage Temperature:Ambient
  • EINECS:247-530-0
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.