Order Entry
Germany
Orders LinkContactUsLinkComponent
 

ADA ≥98%

Glass bottle for solids, 100 g
Catalog # A15267.22
ADA ≥98%
Catalog # A15267.22
CAS Number:  26239-55-4
ADA ≥98%
ADA ≥98%

Glass bottle for solids, 100 g

CAS Number:  26239-55-4
Price
108,00 €
Log in for availability
1 * 100 g
Product Details & Documents

Purity: 98+% CAS No.: 26239-55-4 Molecular Formula: H2NCOCH2N(CH2CO2H)2 Formula Weight: 190.16 Melting Point: ca 218° dec. Beilstein Reference: 1787181 Merck Reference: 14,147 MDL No.: MFCD00008031

Synonyms:  2,2'-[(2-Amino-2-oxoethyl)imino]diacetic acid, N-(2-Acetamido)iminodiacetic acid, 2,2'-((2-Amino-2-oxoethyl)azanediyl)diacetic acid
  • CAS Number:26239-55-4
  • Formula:H₂NC(O)CH₂N(CH₂COOH)₂
  • MDL Number:MFCD00008031
  • Molecular Weight:190.16 g/mol
  • Merck Index:14,147
  • Melting Pt:∼ 218 °C (dec.)
  • Storage Temperature:Ambient
  • EINECS:247-530-0
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications