Orders LinkContactUsLinkComponent
3-Ethoxybenzonitrile 95%
Glass bottle for solids, 25 g
  A13908.14
3-Ethoxybenzonitrile 95%
  A13908.14
 :  25117-75-3
3-Ethoxybenzonitrile 95%
3-Ethoxybenzonitrile 95%
Glass bottle for solids, 25 g
 :  25117-75-3
95,30 €
1 * 25 g
Purity: 95% CAS No.: 25117-75-3 Molecular Formula: C9H9NO Formula Weight: 147.18 Flash Point: 110°(230°F) Refractive Index: 1.529 Boiling Point: 241-244° UN No.: 3276 MDL No.: MFCD00001805
  •  :25117-75-3
  •  :C₉H₉NO
  •  :MFCD00001805
  •  :147.18 g/mol
  •  :241…244 °C
  •  :1.047
  •  :>110 °C (230 °F)
  •  :Ambient
  •  :246-632-2
  •  :3276
  •  :6.1,III
  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g