Order Entry
Germany
Orders LinkContactUsLinkComponent
3-Ethoxybenzonitrile 95%

3-Ethoxybenzonitrile 95%

Glass bottle for solids, 5 g
Catalog # A13908.06
Toxic
Irritant
3-Ethoxybenzonitrile 95%
Catalog # A13908.06
CAS Number:  25117-75-3
3-Ethoxybenzonitrile 95%
3-Ethoxybenzonitrile 95%

Glass bottle for solids, 5 g

CAS Number:  25117-75-3
Price
28,50 €
Log in for availability
1 * 5 g
Product Details & Documents

Purity: 95% CAS No.: 25117-75-3 Molecular Formula: C9H9NO Formula Weight: 147.18 Flash Point: 110°(230°F) Refractive Index: 1.529 Boiling Point: 241-244° UN No.: 3276 MDL No.: MFCD00001805

  • CAS Number:25117-75-3
  • Formula:C₉H₉NO
  • MDL Number:MFCD00001805
  • Molecular Weight:147.18 g/mol
  • Boiling Pt:241…244 °C
  • Density:1.047
  • Flash Pt:>110 °C (230 °F)
  • Storage Temperature:Ambient
  • EINECS:246-632-2
  • UN:3276
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.