Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
BUFFER SOLUTION PH 4 RED AVS TITRINORM 1 * 10 l
Catalog # VWRS32044.410
Supplier:
VWR CUSTOM MADE CHEMICALS
SDS
Expand image
BUFFER SOLUTION PH 4 RED AVS TITRINORM 1 * 10 l
Catalog # VWRS32044.410
Supplier:
VWR CUSTOM MADE CHEMICALS
SDS
Related Information
Documentation
SDS
SDS_EN_UK_VWRS32044.pdf
Documentation