n-Nonane 99%, Reagent Grade
Glass bottle, 500 mL
Supplier: Honeywell Chemicals
n-Nonane 99%, Reagent Grade
Catalog # N29406-500ML
Supplier: Honeywell Chemicals
CAS Number: 111-84-2
Jump to:
Product Details & Documents
- ≥95% for general laboratory use.
- CAS Number:111-84-2
- Formula:H₃C(CH₂)₇CH₃
- MDL Number:MFCD00009574
- Molecular Weight:128.26 g/mol
- Boiling Pt:2.9 %(V)
- Melting Pt:–53 °C
- Melting Pt:-53 °C
- Density:0.72 g/cm³ (20 °C)
- Flash Pt:31 °C
- Storage Temperature:Ambient
- EINECS:203-913-4
- UN:1920
- ADR:3,III
Specifications
Specification Test Results
- Explosion Limit:2.9 %
- Identity (IR):complying
- Assay (GC):Min. 98.5 %
- Refractive index (n 20/D):1.403 - 1.407
- Autoignition Temperature:401 °F
- Vapor Density:4.41 (vs air)
Specifications
- Pack type:Glass bottle
- Pk:500 mL



