n-Nonane 99%, Reagent Grade
Glass bottle, 100 mL
Supplier: Honeywell Chemicals
n-Nonane 99%, Reagent Grade
Catalog # N29406-100ML
Supplier: Honeywell Chemicals
CAS Number: 111-84-2
Jump to:
Product Details & Documents
- ≥95% for general laboratory use.
- CAS Number:111-84-2
- Formula:H₃C(CH₂)₇CH₃
- MDL Number:MFCD00009574
- Molecular Weight:128.26 g/mol
- Boiling Pt:2.9 %(V)
- Melting Pt:–53 °C
- Melting Pt:-53 °C
- Density:0.72 g/cm³ (20 °C)
- Flash Pt:31 °C
- Storage Temperature:Ambient
- EINECS:203-913-4
- UN:1920
- ADR:3,III
Documents
sds_N29406
SDS
Specifications
Specification Test Results
- Explosion Limit:2.9 %
- Identity (IR):complying
- Assay (GC):Min. 98.5 %
- Refractive index (n 20/D):1.403 - 1.407
- Autoignition Temperature:401 °F
- Vapor Density:4.41 (vs air)
Specifications
- Pack type:Glass bottle
- Pk:100 mL
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



