Orders LinkContactUsLinkComponent
Grid Storage Box
Grid Storage Box
Grid storage box
Catalog # SIALG6276-1EA
Supplier:   Merck
Grid Storage Box
Catalog # SIALG6276-1EA
Supplier:  Merck
Grid Storage Box
Grid storage box
Price
48,30 €
Log in for availability
1 * 1 ST
Product Details & Documents
This storage box is made up of plastic.
  • Anti-static
  • Record card is used to record identity of samples
  • Device is anti-static to prevent handling problems due to repulsion or attraction
Specifications
  • Description:
    Grid storage box
  • Seq:
    01