Dulbecco’s phosphate buffered saline (DPBS), Sigma-Aldrich®
DPBS, 100 ml, 100 mL
Catalog # SIALD8537-100ML
Supplier: Merck
Dulbecco’s phosphate buffered saline (DPBS), Sigma-Aldrich®
Catalog # SIALD8537-100ML
Supplier: Merck
CAS Number:
Jump to:
Product Details & Documents
Dulbecco’s phosphate buffered saline is modified with/without calcium chloride and magnesium chloride, liquid, sterile-filtered, suitable for cell culture.
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pk:100 mL
- Description:DPBS, 100 ml
- Cat. No.:SIALD8537-100ML
- M-Clarity quality level:MQ300
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



