Trizmal™ Buffer, Sigma-Aldrich®
Trizmal™ buffer, 1,0 M, 6,3 (concentrate), 50 mL
913-50ML
For use in the Naphthol AS-D Chloroacetate Esterase Kit No. 91-C.
- Trizma® maleate buffer
: Trizmal is a trademark of Sigma-Aldrich Co. LLC
- Pk:50 mL
- Description:Trizmal™ buffer, 1,0 M
- pH value:6,3 (concentrate)
- Cat. No.:913-50ML
- M-Clarity quality level:MQ500
- Storage temperature:2 - 8 °C



