Order Entry
Germany
Orders LinkContactUsLinkComponent
Trizmal™ Buffer, Sigma-Aldrich®

Trizmal™ Buffer, Sigma-Aldrich®

Trizmal™ buffer, 0,2 M, 6,3 (concentrate), 50 mL
Catalog # 903C-50ML
Trizmal™ Buffer, Sigma-Aldrich®
Catalog # 903C-50ML
CAS Number:  
Trizmal™ Buffer, Sigma-Aldrich®

Trizmal™ buffer, 0,2 M, 6,3 (concentrate), 50 mL

CAS Number:  
Price
123,00 €
Log in for availability
1 * 50 mL
Product Details & Documents

For use in the Naphthol AS-D Chloroacetate Esterase Kit No. 91-C.

  • Trizma® maleate buffer

Caution: Trizmal is a trademark of Sigma-Aldrich Co. LLC

Documents

Specifications

Specifications

  • Pk:
    50 mL
  • Description:
    Trizmal™ buffer, 0,2 M
  • pH value:
    6,3 (concentrate)
  • Cat. No.:
    903C-50ML
  • M-Clarity quality level:
    MQ500
  • Storage temperature:
    RT

Specifications