Paraformaldehyde
Glass bottle for solids, 250 g
Catalog # POLS00380-250
Marketsource
Supplier: POLYSCIENCES
Jump to:
Product Details & Documents
Synonyms: Polyoxymethylene
- CAS Number:30525-89-4
- Formula:(CH₂O)n
- MDL Number:MFCD00133991
- Merck Index:13,07096
- Boiling Pt:135…136 °C (13 mmHg)
- Melting Pt:123 °C
- Density:1.46 g/cm³ (20 °C)
- Flash Pt:70 °C
- Storage Temperature:Ambient
- UN:2213
- ADR:4.1,III
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:250 g



