Acrylamide
Glass bottle for solids, 500 g
Catalog # POLS00019-500
Marketsource
Supplier: POLYSCIENCES
Jump to:
Product Details & Documents
Synonyms: Prop-2-enamide
- CAS Number:79-06-1
- Formula:C₃H₅NO
- MDL Number:MFCD00008032
- Molecular Weight:71.08 g/mol
- Boiling Pt:232 °C (1013 hPa)
- Melting Pt:84.5 °C
- Density:1.122 g/cm³ (20 °C)
- Flash Pt:138 °C
- Storage Temperature:Refrigerator
- EINECS:201-173-7
- UN:2074
- ADR:6.1,III
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g



