Paraformaldehyde
Glass bottle for solids, 1 kg
POLS00380-1
Marketsource
: Polyoxymethylene
- :30525-89-4
- :(CH₂O)n
- :MFCD00133991
- :13,07096
- :135…136 °C (13 mmHg)
- :123 °C
- :1.46 g/cm³ (20 °C)
- :70 °C
- :Ambient
- :2213
- :4.1,III
- Pack type:Glass bottle for solids
- Pk:1 kg



