Order Entry
Germany
Orders LinkContactUsLinkComponent
Qualitative filter papers, technical, smooth, grade 3 m/N

Qualitative filter papers, technical, smooth, grade 3 m/N

3 m/N, 385 mm
Catalog # SARTFT-4-305-385
   Low Manufacturing Impact
Supplier:   Sartorius
Qualitative filter papers, technical, smooth, grade 3 m/N
Catalog # SARTFT-4-305-385
Supplier:  Sartorius
   Low Manufacturing Impact
Qualitative filter papers, technical, smooth, grade 3 m/N
Qualitative filter papers, technical, smooth, grade 3 m/N

3 m/N, 385 mm

Price
274,00 €
Log in for availability
1 * 100 ST
Product Details & Documents

These smooth filter papers are made of refined pulp and linters with more than 95% alpha-cellulose content. The ash content of these filter papers is between 0,1 and 0,15%. They are used for routine analyses like clarification, determination of substances, but also as discs with a centre hole for technical applications. Grades with a wet burst resistance >30 kPa are referred to as wet-strengthened and are therefore suitable for pressure or vacuum filtration.

Medium fast rate of filtration, thin, smooth, increased wet strength. Used for acids; general vacuum or pressure filtration applications and less critical analyses; also used to assay the sucrose content in cane sugar solutions with a relatively low viscosity.

Specifications
  • Grade:
    3 m/N
  • Seq:
    12
  • Size:
    385 mm
  • Thickness:
    0,14 mm
  • Cat. No.:
    SARTFT-4-305-385
  • Wet burst strength:
    30 kPa
  • Environmental certification:
    Renewable Energy Certificate
  • Filtration speed:
    30 s
  • Particle retention:
    7 - 10 µm
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Weight:
    65 g/m²