0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
Orders Link
ContactUsLinkComponent
Pens
Permanent Markers
Felt-tip pens
Felt-tip pens
Felt-tip pen, Red
HECH41181004
:
GLASWARENFABRIK KARL HECHT
Felt-tip pens
HECH41181004
:
GLASWARENFABRIK KARL HECHT
Felt-tip pens
Felt-tip pen, Red
2,30 €
1 * 1 ST
-
+
Colour:
Red
Description:
Felt-tip pen
Permanent marker set, dual-tip, Sharpie®
#: HEATHS15094
: HEATHROW
Gaussia luciferase glow assay kit, Pierce™
#: PIER16161
: Thermo Fisher Scientific