ContactUsLinkComponent
Vial, capacity 4ml, colour amber, material glass, 13-425 screw thread, flat bottom, not patched, usable volume 4ml, diameter 15mm, h eight 45mm, residual volume <800µl, NSC silanization reduces loss of polar compounds due to interactions with activ
Catalog # CRMAC4015-S2
Supplier:  CHROMACOL
Vial, capacity 4ml, colour amber, material glass, 13-425 screw thread, flat bottom, not patched, usable volume 4ml, diameter 15mm, h eight 45mm, residual volume <800µl, NSC silanization reduces loss of polar compounds due to interactions with activ
Catalog # CRMAC4015-S2
Supplier:  CHROMACOL