ContactUsLinkComponent
Vial, capacity 2ml, target 10-425 wide opening screw thread, diameter 12mm, height 32mm, flat bottom, material glass, colour clear, usable volume 1.5ml, silanized vial is surface deactivated, target vials feature large, easy to fill opening
Catalog # CRMAC4010-S1W
Supplier:  CHROMACOL
Vial, capacity 2ml, target 10-425 wide opening screw thread, diameter 12mm, height 32mm, flat bottom, material glass, colour clear, usable volume 1.5ml, silanized vial is surface deactivated, target vials feature large, easy to fill opening
Catalog # CRMAC4010-S1W
Supplier:  CHROMACOL