Order Entry
Germany
Orders LinkContactUsLinkComponent
Colour Reference Solutions, Reag.Ph.Eur.

Colour Reference Solutions, Reag.Ph.Eur.

Primary solution blue (1 ml solution contains 62.4 mg CuSO4.5H2O) Reag. Ph. Eur., Glass bottle, 100 mL
Catalog # REGEEPBS01
Supplier:   REAGECON
Colour Reference Solutions, Reag.Ph.Eur.
Catalog # REGEEPBS01
Supplier:  REAGECON
CAS Number:  
Colour Reference Solutions, Reag.Ph.Eur.
Colour Reference Solutions, Reag.Ph.Eur.

Primary solution blue (1 ml solution contains 62.4 mg CuSO4.5H2O) Reag. Ph. Eur., Glass bottle, 100 mL

CAS Number:  
Price
153,00 €
Log in for availability
1 * 100 mL
Product Details & Documents
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Description:
    Primary solution blue (1 ml solution contains 62.4 mg CuSO<sub>4</sub>.5H<sub>2</sub>O) Reag. Ph. Eur.

Specifications