Orders LinkContactUsLinkComponent
HyClone™ CDM4HEK293
HyClone™ CDM4HEK293
CDM4HEK293 medium without phenol red, L-glutamine, HEPES, sodium bicarbonate, Powder, 5 L
Catalog # HYCLSH30859.01
HyClone™ CDM4HEK293
Catalog # HYCLSH30859.01
CAS Number:  
HyClone™ CDM4HEK293
HyClone™ CDM4HEK293
CDM4HEK293 medium without phenol red, L-glutamine, HEPES, sodium bicarbonate, Powder, 5 L
CAS Number:  
Price
423,00 €
Log in for availability
1 * 5 L
Product Details & Documents
HyClone™ CDM4HEK293 media are chemically-defined, animal-derived component free (ADCF) cell culture media.
  • Allows for direct or sequential adaptation
  • Component traceability
  • Manufactured under cGMP
  • Metabolically designed for high cell yield and recombinant protein/vector production
HyClone CDM4HEK293 is a regulatory-friendly medium developed through the Metabolic Pathway Design to support the extremely high productivity and cell density in suspension cultures.
Specifications
Specifications
  • Media Format:
    Powder
  • Pk:
    5 L
  • Description:
    CDM4HEK293 medium without phenol red, L-glutamine, HEPES, sodium bicarbonate