Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Thymolphthalein

Glass bottle for solids, 10 g
Catalog # B23896.09
Thymolphthalein
Catalog # B23896.09
CAS Number:  125-20-2
Thymolphthalein
Thymolphthalein

Glass bottle for solids, 10 g

CAS Number:  125-20-2
Price
27,90 €
Log in for availability
1 * 10 g
Product Details & Documents

CAS No.: 125-20-2 Molecular Formula: C28H30O4 Formula Weight: 430.55 Melting Point: 251-253° Beilstein Reference: 359413 Merck Reference: 14,9401 MDL No.: MFCD00005909

Synonyms:  3,3-Bis(4-hydroxy-5-isopropyl-o-tolyl)phthalide, 3,3-Bis(4-hydroxy-5-isopropyl-2-methylphenyl)isobenzofuran-1(3H)-one
  • CAS Number:125-20-2
  • Formula:C₂₈H₃₀O₄
  • MDL Number:MFCD00005909
  • Molecular Weight:430.54 g/mol
  • Merck Index:14,9401
  • Melting Pt:251…253 °C
  • Storage Temperature:Ambient
  • EINECS:204-729-7
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g

Specifications