Orders LinkContactUsLinkComponent
Supporting frames and ring for reaction vessels
Supporting frames and ring for reaction vessels
Fixed supporting frame, pipe Ø 27 mm, 1800 mm height, for: flat flanges DN 200
Catalog # KGWA7683
Supplier:   KGW
Supporting frames and ring for reaction vessels
Catalog # KGWA7683
Supplier:  KGW
Supporting frames and ring for reaction vessels
Supporting frames and ring for reaction vessels
Fixed supporting frame, pipe Ø 27 mm, 1800 mm height, for: flat flanges DN 200
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    Fixed supporting frame, pipe Ø 27 mm, 1800 mm height, for: flat flanges DN 200