Orders LinkContactUsLinkComponent
Plugs for test tubes
12 mm, Green
Catalog # 212-0527
Supplier:   BOETTGER PAUL
Plugs for test tubes
Catalog # 212-0527
Supplier:  BOETTGER PAUL
Plugs for test tubes
Plugs for test tubes
12 mm, Green
Price
22,10 €
Log in for availability
1 * 1.000 ST
Product Details & Documents
Polyethylene (PE). Ribbed plugs with two sealing ribs, very tight sealing.
  • Not autoclavable
  • Easy handling with a high, ribbed grip
  • Easy to open by tilting 'one-hand operation'
Specifications
  • Colour:
    Green
  • Seq:
    05
  • Size:
    12 mm