pH electrode storage and cleaning solutions, Orion™
Electrode cleaning solution for pH electrodes, single use, 15 ml sachets, 25
Catalog # 662-0783
Supplier: Thermo Orion
pH electrode storage and cleaning solutions, Orion™
Catalog # 662-0783
Supplier: Thermo Orion
CAS Number:
Jump to:
Product Details & Documents
Solutions for storage and cleaning that prolong the working life of pH electrodes. Cleaning solutions are identified by the letters A, B, C, and D; each letter designates a unique formulation.
- Cleaning solution A: for removing protein contaminants
- Cleaning solution B: for removing bacterial contaminants
- Cleaning solution C: for general cleaning
- Cleaning solution D: for removing oil and grease contaminants
Bottles include a small beaker to hold the solution and a plastic pipette to remove the old electrode filling solution.
Storage solution ensures that the pH electrode works properly after prolonged storage, overnight storage and between samples.
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pk:25
- Description:Electrode cleaning solution for pH electrodes, single use, 15 ml sachets
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



